missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Reagent Alcohol, Certified, 70% (v/v), LabChem™

Manufacturer:  LabChem LC222104

 View more versions of this product

Catalog No. LC22210-4



Specifications

Specifications

Reagent Alcohol
56.46,37.63,3.11,2.8
C2H6O
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
Liquid
Certified
Passes Test
Soluble in water
Room Temperature
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Colorless
4L
64-17-5,7732-18-5,67-63-0,67-56-1
C2H6O
UN1987
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.07
70% (v/v)
0.881g/L
Poly Bottle
0.881g/L +/- 0.008
25°C
78°C
-115°C

Chemical Identifiers

64-17-5
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervo

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.<

Danger

EINECSNumber : 200-578-6

Documents
Promotions

Promotions