missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Affymetrix/USB™ Ready-Made Terrific Broth Powder

Provides greater yields of plasmid DNA and recombinant proteins than LB media by allowing for greater cell density

Manufacturer:  Affymetrix/USB™ J7585630

 View more versions of this product

Catalog No. AAJ7585630



Description

Description

  • Off-white free flowing powder
  • pH(5.08%, H2O): 7.2 ± 0.2
Contents
  • Casein Peptone: 12g/L
  • Yeast Extract: 24g/L
  • Potassium Phosphate Dibasic: 12.5g/L
  • Potassium Phosphate Monobasic: 2.3g/L
Specifications

Specifications

Off-white
Plastic Bottle
Terrific Broth
Free Flowing Powder
250g
Provides greater yields of plasmid DNA and recombinant proteins
Documents