missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycine, ≥99%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
$172.77 - $566.42
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.067 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
PubChem CID | 750 |
ChEBI | CHEBI:15428 |
IUPAC Name | 2-aminoacetic acid |
SMILES | C(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AAJ1640736
|
Thermo Fisher Scientific
J1640736 |
500 g | Plastic Bottle |
Each for $172.77
|
|
||||
AAJ16407A1
|
Thermo Fisher Scientific
J16407A1 |
1 kg | Plastic Bottle |
Each for $227.59
|
|
||||
AAJ16407A7
|
Thermo Fisher Scientific
J16407A7 |
5 kg | Plastic Bottle |
Each for $566.42
|
|
||||
Chemical Identifiers
56-40-6 | |
75.067 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
C(C(=O)O)N |
Specifications
∼245°C (decomposition) | |
C2H5NO2 | |
MFCD00008131 | |
Plastic Bottle | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
750 | |
75.07 | |
Molecular Biology | |
500 g | |
Powder |
56-40-6 | |
H2NCH2CO2H | |
635782 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
C(C(=O)O)N | |
75.067 | |
CHEBI:15428 | |
≥99% | |
1.595 | |
1.595 | |
Glycine |
Safety and Handling
EINECSNumber : 200-272-2
RTECSNumber : MB7600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only