missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Affymetrix/USB™ Ready-Made LB Agar Powder

For propagation of bacteria

Manufacturer:  Affymetrix/USB™ J7585130

 View more versions of this product

Catalog No. AAJ7585130



Description

Description

pH(3.5%, H2O): 7.0 ± 0.2
 
Contents:
  • Casein Peptone: 10gm/L
  • Yeast Extract: 5gm/L
  • Sodium Chloride: 5gm/L
  • Agar: 15gm/L
Specifications

Specifications

Free Flowing Powder
250g
Propagation of bacteria
Plastic Bottle
LB Agar Powder
Documents