Please login to your online account to display your discounted pricing

Ready America™ Wall Straps for Carts


Cart-to-wall straps secure carts, metro racks and tubular furniture

Manufacturer: ready america  RWCTWG1

 View more versions of this product

Catalog No. 11-900-036

  • / Each
 

Description & Specifications

Specifications

Product Type Cart-to-Wall Strap™
Color Gray
Quantity Each
Includes One 16 in. strap with D-rings, one 8 in. strap with quick-disconnect buckle, one 0.25 in. Wood stud screw, one 0.25 in. Metal stud toggler with instructions, one 0.25 in. Metal stud screw, one 0.25 x 1 in. washer instructions
For Use With (Application) Secures carts, Metro™ racks, and tubular furniture to studs in walls, with easy disconnect