Please login to your online account to display your discounted pricing

Ready America™ Wall Mount Strap with Side Release Buckle for Medium-Sized Equipment


Secures copiers and other medium-size equipment

Manufacturer: ready america  RW1X48B21

 View more versions of this product

Catalog No. 11-900-029

  • / Each
 

Description & Specifications

Specifications

Color Black
Quantity Each
Includes Two 1 x 48 in. straps using a Steel plate with 3M VHB adhesive and Steel buckle, two 0.25 in. Metal stud toggler with instructions, two 0.25 in. Metal stud screw, two 0.25 in. Wood stud screw, two 0.25 x 1 in. washer, two surface prep pad instructions
For Use With (Application) Secures copiers and other medium size equipment to wall studs with easy disconnect
Product Type Wall Strap™