Please login to your online account to display your discounted pricing

Ready America™ Safety Straps for Filing Cabinets

Ideal for file cabinets, storage cabinets, bookshelves and other similar cabinets not adjacent to a wall

$33.60 - $100.83

Products
Catalog Number Mfr. No. Color Includes Price Quantity    

11-900-024

 
ready america
RG2X14B21
Black 2x14 in. straps w/industrial Velcro hook/loop (2) and prep pads (4) Each for $100.83

11-900-025

 
ready america
RG2X14G21
Gray 2x14 in. straps w/industrial Velcro hook/loop (2) and prep pads (4) Each for $100.83

11-900-031

 
ready america
RW2X12B1
Black 2x12 in. strap w/industrial Velcro hook/loop, 0.25 x 1 in. washer, prep pads, 1 each (0.25 in.) stud toggler, metal screw, wood scre Each for $33.60

11-900-032

 
ready america
RW2X12G1
Gray 2x12 in. strap w/industrial Velcro hook/loop, 0.25 x 1 in. washer, prep pads, 1 each (0.25 in.) stud toggler, metal screw, wood scre Each for $33.60
Description & Specifications

FileSTRAP Kit mounts to cabinet to wall; ganging strap secures cabinets to one another.

FileSTRAP Kit contains:

  • Strap with industrial Velcro™ hook and loop: 2 x 12 in. (5.1 x 30.7cm)
  • Wood stud screws: 0.25 in. (0.64cm)
  • Metal stud togglers with instructions: 0.25 in. (0.64cm)
  • Metal stud screws: 0.25 in. (0.64cm)
  • Washers: 0.25 x 1 in. (0.64x 2.54cm)
  • Surface prep pads
  • Instructions
Ganging File Strap Kit contains:
  • Two straps with industrial Velcro™ hook and loop on each side: 2 x 14 in. (5.1 x 35.6cm)
  • Four surface prep pads
  • Instructions