Please login to your online account to display your discounted pricing

Ready America™ LassoSTRAP Fastener Kits for Microscopes


Secure countertop and desktop equipment during an earthquake

Manufacturer: ready america  RDSG1

 View more versions of this product

Catalog No. 11-900-009

  • / Each
 

Description & Specifications

Specifications

Product Type Lasso strap fastener
Color Gray
Quantity Each
Includes One 1 x 2 in. buckle with 3M VHB adhesive one 0.75 x 30 in. locking strap with D-ring two surface prep pads instructions
For Use With (Equipment) Countertop and desktop pedestal equipment weighing less than 25 lb.