Please login to your online account to display your discounted pricing

Ready America™ HPLC Stack Fastener Kits


Secures HPLC stacks

Manufacturer: ready america  RDHP3G1

 View more versions of this product

Catalog No. 11-900-003

  • / Each
 

Description & Specifications

Specifications

Product Type HPLC 3 stack fastener kit
Color Gray
Quantity Each
Includes Two 2 x 2 in. countertop buckles with adhesive, six 1 x 2 in. equipment buckles with adhesive, two 12 in. coated locking straps, 6 prep pads
For Use With (Application) Secure HPLC with base buckles to countertop and equipment buckles to 2 stacked components on both sides