Ready America™ HPLC Stack Fastener Kits
Secures HPLC stacks
Manufacturer:  Ready America RDHP6G1
 View more versions of this product
Catalog No. 11-900-006
Specifications Description & Specifications
Specifications
Gray
Two 2 x 2 in. countertop buckles with adhesive, twelve 1 x 2 in. equipment buckles with adhesive, two 12 in. coated locking straps, 12 prep pads
Each
Secure HPLC with base buckles to countertop and equipment buckles to 2 stacked components on both sides
HPLC 6 stack fastener kit