Please login to your online account to display your discounted pricing

Ready America™ Floor-Mounting Safety Fasteners for Large Equipment


Ideal for multi-door refrigerators, freezers, shakers, incubators and other large floor equipment

Manufacturer: ready america  RFR30G1

 View more versions of this product

Catalog No. 11-900-020

  • / Each
 

Description & Specifications

Specifications

Product Type Ratchet strap
For Use With (Equipment) Multi door refrigerators, freezers, shakers, incubators and other large floor equipment
Includes One 2 x 30 in. Strap with wide handle ratchet/floor mount plate, one 2 x 180 in. strap with floor mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, instructions
Quantity Each
Color Gray