Please login to your online account to display your discounted pricing

Ready America™ Floor-Mounting Safety Fasteners for Large Equipment


Ideal for multi-door refrigerators, freezers, shakers, incubators and other large floor equipment

Manufacturer: ready america  RFCB6H1

 View more versions of this product

Catalog No. 11-900-018

  • / Each
 

Description & Specifications

Specifications

Product Type Corner Bracket
Material Stainless Steel
Color Silver
Quantity Each
Includes Round or square legs as well as corners with legs 6 H x 0.25 in. thick, two each 0.37 in. drop anchors, 0.37 in. anchor bolts, and 0.37 in. washers
For Use With (Equipment) Multi door refrigerators, freezers, shakers, incubators and other large floor equipment