Ready America™ Floor-Mounting Safety Fasteners for Large Equipment
Ideal for multi-door refrigerators, freezers, shakers, incubators and other large floor equipment
$223.20 - $321.06
Specifications
For Use With (Equipment) Multi door refrigerators, freezers, shakers, incubators and other large floor equipment
Quantity Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Color Product Type Price Quantity  
Catalog Number Mfr. No. Includes Color Product Type Price Quantity  
11-900-018 Ready America
RFCB6H1
Round or square legs as well as corners with legs 6 H x 0.25 in. thick, two each 0.37 in. drop anchors, 0.37 in. anchor bolts, and 0.37 in. washers Silver Corner Bracket Each for $321.06
11-900-022 Ready America
RFR90CBG1
One 2 x 90 in. Strap with wide handle ratchet/floor mount plate, one 2 x 180 in. strap with floor mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, two corner brackets, instructions Gray Ratchet strap Each for $281.57
11-900-019 Ready America
RFR180G1
One 2 x 180 in. Strap with wide-handle ratchet/floor-mount plate, one 2 x 180 in. strap with floor-mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, instructions Gray Ratchet strap Each for $274.69
11-900-021 Ready America
RFR60G1
One 2 x 60 in. Strap with wide handle ratchet/floor mount plate, one 2 x 180 in. strap with floor mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, instructions Gray Ratchet strap Each for $233.49
11-900-020 Ready America
RFR30G1
One 2 x 30 in. Strap with wide handle ratchet/floor mount plate, one 2 x 180 in. strap with floor mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, instructions Gray Ratchet strap Each for $223.20
11-900-023 Ready America
RFR90G1
One 2 x 90 in. Strap with wide handle ratchet/floor mount plate, one 2 x 180 in. strap with floor mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, instructions Gray Ratchet strap Each for $240.37
Specifications
Description
Ratchet Strap Kits are ideal for multi-door refrigerators, freezers, shakers, incubators and other large floor equipment. Corner Bracket Kit works for equipment with round or square legs as well as corners without legs.
Corner Bracket Kit contains:
  • Stainless steel corner bracket: 0.25 x 6 in.
  • Two each drop anchors, anchor bolts and washers: 0.38 in. (0.95cm)
  • Instructions
All Ratchet Strap Kits contain:
  • 2 x 180 in. (5.1cm x 4.6m) strap with floor-mounting plate
  • Two each drop anchors, anchor bolts and washers: 0.38 in. (0.95cm)
  • Instructions
  • Each kit contains one strap with wide-handle ratchet and floor-mounting plate:
    • 30 in. Kit: 2 x 30 in. (76.2cm) strap
    • 60 in. Kit: 2 x 60 in. (15.2m) strap
    • 90 in. Kit: 2 x 90 in. (22.9m) strap
    • 180 in. Kit: 2 x 180 in. (45.7cm) strap
Description & Specifications
Specifications
Multi door refrigerators, freezers, shakers, incubators and other large floor equipment
Each