Please login to your online account to display your discounted pricing

Ready America™ Floor-Mounting Safety Fasteners for Large Equipment


Ideal for multi-door refrigerators, freezers, shakers, incubators and other large floor equipment

Manufacturer: ready america  RFR180G1

 View more versions of this product

Catalog No. 11-900-019

  • / Each
 

Description & Specifications

Specifications

Product Type Ratchet strap
Color Gray
Quantity Each
Includes One 2 x 180 in. Strap with wide-handle ratchet/floor-mount plate, one 2 x 180 in. strap with floor-mount plate, two 0.37 in. drop anchors, two 0.37 in. anchor bolts, two 0.37 in. washers, instructions
For Use With (Equipment) Multi door refrigerators, freezers, shakers, incubators and other large floor equipment