Please login to your online account to display your discounted pricing

Ready America™ Floor-Mounting High Z-Bracket for Equipment with Adjustable Legs


Secures equipment with adjustable legs in the event of an earthquake

Manufacturer: ready america  RZ1001

 View more versions of this product

Catalog No. 11-900-041

  • / Each
 

Description & Specifications

Specifications

Product Type Bracket
Material Stainless Steel
Color Silver
Quantity Each
Includes Z-bracket with 2 in. high rise, 0.375 in. drop anchors, 0.375 in. anchor bolts, 0.375 in. washers
For Use With (Application) Secures equipment with adjustable legs