missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ready America™ Floor-Mounting High Z-Bracket for Equipment with Adjustable Legs

Secures equipment with adjustable legs in the event of an earthquake

$171.69 - $206.03

Specifications

Color Silver
Material Stainless Steel
Product Type Bracket
For Use With (Application) Secures equipment with adjustable legs
Quantity Each
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
11-900-040 Ready America
RZ0501
Z-bracket with 1 in. high rise, 0.375 in. drop anchors, 0.375 in. anchor bolts, 0.375 in. washers Each for $171.69
11-900-041 Ready America
RZ1001
Z-bracket with 2 in. high rise, 0.375 in. drop anchors, 0.375 in. anchor bolts, 0.375 in. washers Each for $206.03
Specifications

Description

Kit contains:

  • 1 or 2 in. (2.54/5.1cm) Z-Bracket
  • Two each drop anchors, anchor bolts and washers: 0.38 in. (0.95cm)

Description & Specifications

Specifications

Silver
Bracket
Each
Stainless Steel
Secures equipment with adjustable legs