Please login to your online account to display your discounted pricing

Ready America™ Fastener Kits for Countertop Equipment


Used for countertop and desktop equipment

Manufacturer: ready america  RDT64W1

 View more versions of this product

Catalog No. 11-900-017

  • / Each
 

Description & Specifications

Specifications

Product Type Equipment Fastener
Quantity Each
Color White
For Use With (Equipment) Countertop and desktop equipment that weigh less than 200 lb.
Includes Four 2 x 3 in. buckles with 3M VHB adhesive, two 1 x 8 in. locking straps and two surface prep pads, instructions