missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Reading Glass Magnifiers

Three sizes for quick, convenient magnification.

Manufacturer:  Eisco™ PH0510F

 View more versions of this product

Catalog No. S95954



Description

Description

  • Chrome-plated metal frames with plastic handles
  • Packed in cardboard box

Specifications

Specifications

2.4 in.
7.4 x 6.2 x 3.14 in. (19 x 16 x 8cm)
3.14 in.
Chrome plated bakelite handle
7.4 in.
19cm
1kg
16cm
Glass
6.09cm
For quick, convenient magnification
8cm
0.5 in.
1.27cm
2.2 lb.
6.2 in.
2.25x
Reading Magnifier
SDS
Documents
Product Certifications
Promotions

Promotions