missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Reading Glass Magnifiers
Three sizes for quick, convenient magnification.
Manufacturer: Eisco™ PH0510B
Description
- Chrome-plated metal frames with plastic handles
- Packed in cardboard box
Specifications
2 in. | |
6.2 x 6.2 x 2.75 in. (16 x 16 x 7cm) | |
2.75 in. | |
Chrome plated bakelite handle | |
6.2 in. | |
16cm | |
0.67kg | |
16cm | |
Glass |
5.08cm | |
For quick, convenient magnification | |
7cm | |
7.9 in. | |
20cm | |
1.47 lb. | |
6.2 in. | |
3x | |
Reading Magnifier |