RCI Conductivity Indicators
Ideal for comparing various solutions across a wide range of conductivity levels.
Manufacturer: Fisher Scientific RCIDX
Requires : 9V transistor battery
Description
Simply immerse the probes in the solution and record the display lamp lit. The higher the numbered display lamp lit, the greater the solution conductivity.
- Unique, multiple lamp conductivity display determines the relative conductivity of solutions
- Choose from the 10-level RCI Junior or the advanced, 20-level RCI-DX conductivity indicator
Specifications
LED | |
RCI-DX (20-level RCI advanced) | |
9V |
Ideal for comparing various solutions across a wide range of conductivity levels | |
Conductivity |