missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Razor Blades

Single-edged surgical carbon-steel blades are easily and safely gripped.

Manufacturer:  Fisher Scientific 0140CS

 View more versions of this product

Catalog No. S17302




Description

Description

  • Dimensions: 1.5L × 0.75 in.W (3.8 × 1.9cm)

Order Info

Individually shelled in packs of 100.

Specifications

Specifications

1.5 in.
Razor Blade
0.2mm (0.009 in.)
3.81cm
Carbon Steel
SDS
Documents
Product Certifications