Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Rat TNF alpha ELISA Kits


Measure levels of rat tumor necrosis factor alpha (TNFa) levels in rat serum, plasma and culture supernatant samples using 96-well plates.

Manufacturer: thermo scientific™ pierce™  ER3TNFA2

 View more versions of this product

Catalog No. ENER3TNFA2

  • / Each
 

Description & Specifications

Specifications

Specificity No significant cross-reaction with human TNFα, human TNFβ or porcine TNFα. Mouse TNFα cross-reacts weakly (< 3%)
Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <15pg/mL
Kit Contents 30X Wash Buffer, 2 x 50mL
Sufficient For 2 x 96 microplate wells
Description Rat TNF alpha ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 2 plate kit
Total Assay Time 3 hr.
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Formulation Complete ELISA kit with coated 96-well plates
Target Natural and recombinant rat TNFa (rat tumor necrosis factor alpha)
Sample Size 50µL
Assay Range 31 to 2500pg/mL

Assay TNFα (rat tumor necrosis factor alpha) levels in rat serum, plasma (EDTA, heparin) and culture supernatant samples with our Thermo Scientific Rat TNF alpha ELISA Kits.

These rat TNF alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Natural and recombinant rat TNFα (rat tumor necrosis factor alpha)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 31 to 2500pg/mL

  • Sensitivity: < 15pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human TNFα, human TNFβ or porcine TNFα. Mouse TNFα cross-reacts weakly (< 3%)

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.