missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Rat TGF-beta1 Platinum ELISA Kit

For the quantitative detection of rat TGF-beta1.

Manufacturer:  Invitrogen™ LSBMS6233TEN

 View more versions of this product

Catalog No. 501125467



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat TGF-beta1
Biotin-Conjugate anti-rat TGF-beta1 polyclonal antibody
Streptavidin-HRP
Rat TGF-beta1 Standard lyophilized, 4000pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrat Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

ELISA
12pg/mL
100μL (pretreated)
tgf beta 1
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
Rat
Camurati-Engelmann disease, DPD1
Documents