missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Rat MCP-1 ELISA Kits

Measure levels of rat monocyte chemoattractant protein-1 (MCP1) in rat serum, plasma and cell culture supernatant samples.

$651.05 - $1,145.50

Specifications

Assay Range 38 to 1500pg/mL
Description Rat MCP-1 ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ENERMCP1 Thermo Scientific™ Pierce™
PIERMCP1
1 plate kit Each for $651.05
ENERMCP12 Thermo Scientific™ Pierce™
ERMCP12
2 plate kit Each for $1,145.50
Specifications

Description

Assay MCP-1 (rat monocyte chemoattractant protein-1) levels in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Rat MCP-1 ELISA Kits.

These rat MCP-1 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of monocyte chemoattractant protein-1 (MCP-1) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat MCP-1 (monocyte chemoattractant protein-1)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 38 to 1500pg/mL

  • Sensitivity: < 5pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IFNγ, IL-1α, IL-1β, IL-2, IL-4, IL-6, IL-10, MIP-1α, RANTES, mouse MCP-1, or human MCP-1

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Description & Specifications

Specifications

38 to 1500pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV <10%; inter-assay CV <10%
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with rat IFNg, IL-1a, IL-1ß, IL-2, IL-4, IL-6, IL-10, MIP-1a, RANTES, mouse MCP-1, or human MCP-1
3 hr.
Rat MCP-1 ELISA Kit
Absorbance at 450nm minus absorbance at 550nm
50.0µL
<5pg/mL
Natural and recombinant rat MCP-1 (monocyte chemoattractant protein-1)