Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Rat MCP-1 ELISA Kits


Measure levels of rat monocyte chemoattractant protein-1 (MCP1) in rat serum, plasma and cell culture supernatant samples.

Manufacturer: thermo scientific™ pierce™  PIERMCP1

 View more versions of this product

Catalog No. ENERMCP1

  • / Each
 

Description & Specifications

Specifications

Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <5pg/mL
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Formulation Complete ELISA kit with coated 96-well plate
Kit Contents 30X Wash Buffer, 50mL
Assay Range 38 to 1500pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 1 plate kit
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50.0µL
Specificity No significant cross-reaction with rat IFNg, IL-1a, IL-1ß, IL-2, IL-4, IL-6, IL-10, MIP-1a, RANTES, mouse MCP-1, or human MCP-1
Sufficient For 96 microplate wells
Total Assay Time 3 hr.
Target Natural and recombinant rat MCP-1 (monocyte chemoattractant protein-1)
Description Rat MCP-1 ELISA Kit

Assay MCP-1 (rat monocyte chemoattractant protein-1) levels in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Rat MCP-1 ELISA Kits.

These rat MCP-1 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of monocyte chemoattractant protein-1 (MCP-1) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Natural and recombinant rat MCP-1 (monocyte chemoattractant protein-1)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 38 to 1500pg/mL

  • Sensitivity: < 5pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IFNγ, IL-1α, IL-1β, IL-2, IL-4, IL-6, IL-10, MIP-1α, RANTES, mouse MCP-1, or human MCP-1

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.