missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Rat IL-6 ELISA Kits

Measure levels of rat interleukin-6 in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates.

$2,436.00 - $2,436.00

Specifications

Assay Range 62.5 to 4000pg/mL
Description Rat IL-6 ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Dependent upon sample type
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIER3IL65 Thermo Scientific™ Pierce™
ER3IL65
5 plate kit Each for $2,436.00
Specifications

Description

Assay IL-6 (rat interleukin-6) levels in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific™ Rat IL-6 ELISA Kits.

These rat IL-6 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat interleukin-6 (IL-6)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 62.5 to 4000pg/mL

  • Sensitivity: < 16pg/mL

  • Precision: Dependent upon sample type

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IL-10 and TNFα; mouse IL-6, IL-1β, TNFα, IL-4, IL-2 and IL-1α; or human IL-8 and GM-CSF

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Description & Specifications

Specifications

62.5 to 4000pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Dependent upon sample type
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with rat IL-10 and TNFa; mouse IL-6, IL-1ß, TNFa, IL-4, IL-2 and IL-1a; or human IL-8 and GM-CSF
4 hr.
Rat IL-6 ELISA Kit
Absorbance at 450nm minus absorbance at 550nm
50µL
<16pg/mL
Natural and recombinant rat interleukin-6 (IL-6)