Please login to your online account to display your discounted pricing

eBioscience Rat IL-4 Platinum ELISA Kit

For the quantitative detection of rat IL-4.

$651.05 - $3,116.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym B-cell stimulatory factor 1, B cell growth factor 1
Target IL-4
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-4
Biotin-Conjugate anti-rat IL-4 monoclonal antibody
Streptavidin-HRP
Rat IL-4 Standard lyophilized, 200pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018354

 
ebioscience
LSBMS628
96 tests Each for $651.05

5018356

 
ebioscience
LSBMS628TEN
10 x 96 tests Each for $3,116.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym B-cell stimulatory factor 1, B cell growth factor 1
Target IL-4
Conjugate Biotin
Sample Type Cell culture supernatant, serum
Sensitivity 0.2pg/mL
Species Rat

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency