Please login to your online account to display your discounted pricing

eBiosciencetrade; Rat IL-17A (homodimer) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of rat IL-17A homodimer protein levels from samples including serum and cell culture supernatants.

$332.05 - $694.55

Specifications

For Use With (Application) ELISA
Synonym Interleukin-17A, IL-17AA
Target IL-17A
Sensitivity 2pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

 Alert

Contains Azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017274

 
ebioscience&trade
LS88717088
Each for $665.55

501128810

 
ebioscience
LS88717086
96-well plates 10 x 96 tests Each for $694.55

501128951

 
ebioscience&trade
LS88717022
Each for $332.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-17A, IL-17AA
Target IL-17A
Sensitivity 2pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Rat

Rat IL-17A ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)