missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat IL-10 Quantikine ELISA Kit, R&D Systems

a 4.5 hour solid phase ELISA designed to measure rat IL-10 levels in cell culture supernates, serum, and plasma.

$624.40 - $2,996.88

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 25325
Sample Type Cell Culture Supernates, Serum, EDTA Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
R1000 R & D Systems
RDSR1000
1 Plate Each for $624.40
SR1000 R & D Systems
RDSSR1000
6 Plates Each for $2,996.88
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL)
Sensitivity: 10pg/mL
Specificity: Natural and recombinant rat IL-10
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
25325
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL)
Rat
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum, EDTA Plasma
10pg/mL
Natural and recombinant rat IL-10