missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Rat IL-10 Platinum ELISA Kit

For the quantitative detection of rat IL-10.

Manufacturer:  Invitrogen™ LSBMS629TWO

 View more versions of this product

Catalog No. 501125470



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-10
Biotin-Conjugate anti-rat IL-10 monoclonal antibody
Streptavidin-HRP
Rat IL-10 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

Biotin
2 x 96 tests
Cell culture supernatant, serum
Rat
TGIF, MCSF III, CSIF, B TCGF
ELISA
1.5pg/mL
50μL
IL-10
Documents

For Research Use Only