missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Rat IL-10 Platinum ELISA Kit
For the quantitative detection of rat IL-10.
Manufacturer: Invitrogen™ LSBMS629FIVE
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-10
Biotin-Conjugate anti-rat IL-10 monoclonal antibody
Streptavidin-HRP
Rat IL-10 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
Biotin | |
5 x 96 tests | |
Cell culture supernatant, serum | |
Rat | |
TGIF, MCSF III, CSIF, B TCGF |
ELISA | |
1.5pg/mL | |
50μL | |
IL-10 |
For Research Use Only