missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Rat IL-10 ELISA Kits

Measure levels of rat interleukin-10 in rat serum, plasma (EDTA, heparin, sodium citrate) and cell culture supernatant samples using 96-well plates.

$651.05 - $1,724.10

Specifications

Assay Range 16 to 500pg/mL
Description Rat IL-10 ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Dependent upon sample type (generally less than 10% CV)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ENERIL10 Thermo Scientific™ Pierce™
PIERIL10
1 plate kit Each for $651.05
ENERIL102 Thermo Scientific™ Pierce™
ERIL102
2 plate kit Each for $1,724.10
Specifications

Description

Assay IL-10 (rat interleukin-10) levels in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific™ Rat IL-10 ELISA Kits.

These rat IL-10 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-10 (IL-10) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution.The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat interleukin-10 (IL-10)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 16 to 500pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Dependent upon sample type (generally less than 10% CV)

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IFNγ, IL-1α, IL-1β, IL-2, IL-4, IL-6, MCP-1, MIP-1α, RANTES, TNFα, human IL-10, or mouse IL-10

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Description & Specifications

Specifications

16 to 500pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Dependent upon sample type (generally less than 10% CV)
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with rat IFNg, IL-1a, IL-1ß, IL-2, IL-4, IL-6, MCP-1, MIP-1a, RANTES, TNFa, human IL-10, or mouse IL-10
3 hr.
Rat IL-10 ELISA Kit
Absorbance at 450nm minus absorbance at 550nm
50µL
<3pg/mL
Natural and recombinant rat interleukin-10 (IL-10)