Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Rat IL-10 ELISA Kits


Measure levels of rat interleukin-10 in rat serum, plasma (EDTA, heparin, sodium citrate) and cell culture supernatant samples using 96-well plates.

Manufacturer: thermo scientific™ pierce™  ERIL102

 View more versions of this product

Catalog No. ENERIL102

  • / Each
 

Description & Specifications

Specifications

Sensitivity <3pg/mL
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Kit Contents 30X Wash Buffer, 2 x 50mL
Description Rat IL-10 ELISA Kit
Specificity No significant cross-reaction with rat IFNg, IL-1a, IL-1ß, IL-2, IL-4, IL-6, MCP-1, MIP-1a, RANTES, TNFa, human IL-10, or mouse IL-10
Formulation Complete ELISA kit with coated 96-well plates
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50µL
Precision Dependent upon sample type (generally less than 10% CV)
Sufficient For 2 x 96 microplate wells
Assay Range 16 to 500pg/mL
Target Natural and recombinant rat interleukin-10 (IL-10)
Total Assay Time 3 hr.
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 2 plate kit

Assay IL-10 (rat interleukin-10) levels in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific™ Rat IL-10 ELISA Kits.

These rat IL-10 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-10 (IL-10) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution.The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Natural and recombinant rat interleukin-10 (IL-10)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 16 to 500pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Dependent upon sample type (generally less than 10% CV)

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IFNγ, IL-1α, IL-1β, IL-2, IL-4, IL-6, MCP-1, MIP-1α, RANTES, TNFα, human IL-10, or mouse IL-10

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.