Please login to your online account to display your discounted pricing

Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSRLB00

 View more versions of this product

Catalog No. RLB00

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 2 Plates
Sample Type Cell Culture Supernates, Serum, EDTA Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (17μL), EDTA Plasma (17μL)
Sensitivity 5pg/mL
Sufficient For Sufficient materials to run ELISAs on at least two 96-well plates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Quantity 1 Kit
Species Rat
Gene ID (Entrez) 24494
Product Type Solid Phase Sandwich ELISA
Specificity Natural and recombinant rat IL-1 beta