Please login to your online account to display your discounted pricing

Rat IgM Kappa Light Chain, Clone: RGM, FITC, Isotype control, Novus Biologicals™

$219.00 - $736.00

Specifications

Antigen IgM Kappa Llight Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14395801

 
novus biologicals canada
NBP14395801MG
0.1mg Each for $329.00

NBP14395805

 
novus biologicals canada
NBP143958005MG
0.05mg Each for $219.00

NBP1439585

 
novus biologicals canada
NBP14395805MG
0.5mg Each for $736.00
Description & Specifications

Specifications

Antigen IgM Kappa Llight Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.