Invitrogen™ eBioscience™ Rat IgM ELISA Ready-SET-Go!™ Kit
For quantitative detection of rat IgM.
Manufacturer:  Invitrogen™ LS885054077
 View more versions of this product
Catalog No. 501125144
Includes: Capture Antibody Concentrate
Description
Description
Rat IgM Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
ELISA
0.3125ng/mL
10μL (prediluted 1:10,000)
2° to 8°C
Immunoglobulin M
20 x 96 tests
Cell culture supernatant, serum and plasma
Rat
IgM