missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Rat IgM ELISA Ready-SET-Go!™ Kit
For quantitative detection of rat IgM.
Manufacturer: Invitrogen™ LS885054086
Includes: Capture Antibody Concentrate
Description
Rat IgM Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
ELISA | |
10 x 96 tests | |
Cell culture supernatant, serum and plasma | |
Rat | |
IgM |
96-well plates | |
0.3125ng/mL | |
10μL (prediluted 1:10,000) | |
2° to 8°C | |
Immunoglobulin M |
For Research Use Only