Please login to your online account to display your discounted pricing

Rat IgG2b Kappa Light Chain, Clone: 149/10H5, Unlabeled, Isotype control, Novus Biologicals™

$175.00 - $285.00

Specifications

Antigen IgG2b Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Unlabeled
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14332305

 
novus biologicals canada
NBP143323005MG
0.05mg Each for $175.00

NBP1433235

 
novus biologicals canada
NBP14332305MG
0.5mg Each for $285.00
Description & Specifications

Specifications

Antigen IgG2b Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Unlabeled
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.