Please login to your online account to display your discounted pricing

Rat IgG2b Kappa Light Chain, Clone: 149/10H5, Unlabeled, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143323005MG

 View more versions of this product

Catalog No. NBP14332305

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Primary or Secondary Primary
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Isotype IgG2b Kappa
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.05mg
Monoclonal or Polyclonal Monoclonal
Antigen IgG2b Kappa Light Chain
Host Species Rat
Format Liquid
Conjugate Unlabeled
Test Specificity The monoclonal rat IgG2b, kappa immunoglobulin is useful as an isotype control for rat IgG2b antibodies.
Concentration 0.5mg/mL
Clone 149/10H5