missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat IgG2b Kappa Light Chain, Clone: 149/10H5, FITC, Isotype control, Novus Biologicals™

$219.00 - $736.00

Specifications

Antigen IgG2b Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications Flow Cytometry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1439625 Novus Biologicals Canada
NBP14396205MG
0.5mg Each for $736.00
NBP1439625M Novus Biologicals Canada
NBP143962005MG
0.05mg Each for $219.00
Specifications Description & Specifications

Specifications

IgG2b Kappa Light Chain
Rat
Flow Cytometry
Primary
FITC
Store at 4°C in the dark.