Please login to your online account to display your discounted pricing

Rat IgG2a Kappa Light Chain, Clone: R2a, Unlabeled, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14332105MG

 View more versions of this product

Catalog No. NBP1433215

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Format Liquid
Host Species Rat
Quantity 0.5mg
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Conjugate Unlabeled
Antigen IgG2a Kappa Light Chain
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Isotype IgG2a Kappa
Monoclonal or Polyclonal Monoclonal
Test Specificity The monoclonal rat IgG2a, kappa immunoglobulin is useful as an isotype control for rat IgG2a antibodies.
Clone R2a