missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat IgG2a Kappa Light Chain, Clone: R2a, PE-Cy7, Isotype control, Novus Biologicals™

$252.00 - $373.00

Specifications

Antigen IgG2a Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate PE-Cy7
Applications Flow Cytometry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14294501 Novus Biologicals Canada
NBP14294501MG
0.1mg Each for $373.00
NBP14294505 Novus Biologicals Canada
NBP142945005MG
0.05mg Each for $252.00
Specifications Description & Specifications

Specifications

IgG2a Kappa Light Chain
Rat
Flow Cytometry
Primary
PE-Cy7
Store at 4°C. Do not freeze.