Please login to your online account to display your discounted pricing

Rat IgG2a Kappa Light Chain, Clone: R2a, FITC, Isotype control, Novus Biologicals™

$219.00 - $758.00

Specifications

Antigen IgG2a Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14395901

 
novus biologicals canada
NBP14395901MG
0.1mg Each for $329.00

NBP1439595

 
novus biologicals canada
NBP14395905MG
0.5mg Each for $758.00

NBP14395905

 
novus biologicals canada
NBP143959005MG
0.05mg Each for $219.00
Description & Specifications

Specifications

Antigen IgG2a Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications Flow Cytometry
Storage Requirements Store at 4°C in the dark.