missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat IgG2a Kappa Light Chain, Clone: R2a, Biotin, Isotype control, Novus Biologicals™

$219.00 - $736.00

Specifications

Antigen IgG2a Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Biotin
Applications Flow Cytometry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14356101 Novus Biologicals Canada
NBP14356101MG
0.1mg Each for $329.00
NBP1435615 Novus Biologicals Canada
NBP14356105MG
0.5mg Each for $736.00
NBP14356105 Novus Biologicals Canada
NBP143561005MG
0.05mg Each for $219.00
Specifications Description & Specifications

Specifications

IgG2a Kappa Light Chain
Rat
Flow Cytometry
Primary
Biotin
Store at 4°C. Do not freeze.