missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Rat IgG2a ELISA Ready-SET-Go!™ Kit
For quantitative detection of rat IgG2a.
Manufacturer: Invitrogen™ LS885051086
Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Rat IgG2a isotype control (standard), lyophilized, 50ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Description
Rat IgG2a Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
ELISA | |
10 x 96 tests | |
Serum, plasma and cell culture supernatant | |
Rat | |
IgG2a |
96-well plates | |
0.39ng/mL | |
10μL (prediluted 1:100,000) | |
2° to 8°C | |
Immunoglobulin G2a |
For Research Use Only