missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Rat IgG2a ELISA Ready-SET-Go!™ Kit

For quantitative detection of rat IgG2a.

Manufacturer:  Invitrogen™ LS885051076

 View more versions of this product

Catalog No. 501125137



Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Rat IgG2a isotype control (standard), lyophilized, 50ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)

Description

Description

Rat IgG2a Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
20 x 96 tests
Serum, plasma and cell culture supernatant
Rat
IgG2a
96-well plates
0.39ng/mL
10μL (prediluted 1:100,000)
2° to 8°C
Immunoglobulin G2a
Documents

For Research Use Only