missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat IgG1 Kappa Light Chain, Clone: RG1, Unlabeled, Isotype control, Novus Biologicals™

$175.00 - $285.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Unlabeled
Applications ELISA,Flow Cytometry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1433225 Novus Biologicals Canada
NBP14332205MG
0.5mg Each for $285.00
NBP1433225M Novus Biologicals Canada
NBP143322005MG
0.05mg Each for $175.00
Specifications Description & Specifications

Specifications

IgG1 Kappa Light Chain
Rat
ELISA,Flow Cytometry
Primary
Unlabeled
Store at 4°C. Do not freeze.