Please login to your online account to display your discounted pricing

Rat IgG1 Kappa Light Chain, Clone: RG1, Unlabeled, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14332205MG

 View more versions of this product

Catalog No. NBP1433225

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Host Species Rat
Quantity 0.5mg
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Test Specificity The monoclonal rat IgG1, kappa immunoglobulin is useful as an isotype control for rat IgG1 antibodies.
Isotype IgG1 Kappa
Format Liquid
Conjugate Unlabeled
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Clone RG1
Concentration 0.5mg/mL
Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary