Please login to your online account to display your discounted pricing

Rat IgG1 Kappa Light Chain, Clone: RG1, PE-Cy7, Isotype control, Novus Biologicals™

$252.00 - $373.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate PE-Cy7
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14294601

 
novus biologicals canada
NBP14294601MG
0.1mg Each for $373.00

NBP14294605

 
novus biologicals canada
NBP142946005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate PE-Cy7
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.