Please login to your online account to display your discounted pricing

Rat IgG1 Kappa Light Chain, Clone: RG1, FITC, Isotype control, Novus Biologicals™

$219.00 - $758.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications ELISA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14396001

 
novus biologicals canada
NBP14396001MG
0.1mg Each for $329.00

NBP14396005

 
novus biologicals canada
NBP143960005MG
0.05mg Each for $219.00

NBP1439605

 
novus biologicals canada
NBP14396005MG
0.5mg Each for $758.00
Description & Specifications

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate FITC
Applications ELISA
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.